Ethylene glycol 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

14675-0010 14675-0250 146750025
TS14675-0010EA 126 USD
TS14675-0010 TS14675-0025 TS14675-0250 TS146750025
Ethylene glycol 99+%, Extra Pure
Ethylene glycol
Formula: HOCH₂CH₂OH
Boiling Pt: 196…198 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.113 g/cm³
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4330 (20°C, 589 nm)
Water ≤0.1 % (Coulometric)
Sulfated ash ≤0.05 %
GC ≥99.0 %
Color scale ≤10 APHA
Acidity (CH3COOH) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR