2-Propanol, extra dry 99.8%, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
32696-0010
32696-0025
326961000
TS32696-0010EA
93
USD
TS32696-0010
TS32696-0025
TS326961000
2-Propanol, extra dry 99.8%, AcroSeal™
2-Propanol
Formula:
(CH₃)₂CHOH Boiling Pt: 81…83 °C (760 mmHg) Melting Pt: –89.5 °C Density: 0.785 g/cm³ Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II REACH: 01-2119457558-25 Merck Index: 13,05228 |
|
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % (Coulometric) |
GC | ≥99.75 % |
Residue after evaporation | ≤0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...