1-Octanol 99%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
n-Octanol
15063-0025
15063-0010
15063-0250
15063-2500
TS15063-0025EA
188
USD
TS15063-0025
TS15063-0010
TS15063-0250
TS15063-2500
1-Octanol 99%, pure
1-Octanol
Formula:
H₃C(CH₂)₆CH₂OH MW: 130.23 g/mol Boiling Pt: 195 °C (760 mmHg) Melting Pt: –16 °C Density: 0.824 g/cm³ Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002988 CAS Number: 111-87-5 EINECS: 203-917-6 Merck Index: 13,06784 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4280 to 1.4310 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...