1-Decanol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Hydroxy decane, 1-Decyl alcohol, Capric alcohol, Decan-1-ol, Nonyl carbinol, Decyl alcohol

16576-0025 16576-0010 16576-0100
TS16576-0025EA 118 USD
TS16576-0025 TS16576-0010 TS16576-0100
1-Decanol 98+%
1-Decanol
Formula: H₃C(CH₂)₈CH₂OH
MW: 158.28 g/mol
Boiling Pt: 220…240 °C (760 mmHg)
Melting Pt: 6 °C
Density: 0.829 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00004747
CAS Number: 112-30-1
EINECS: 203-956-9
Merck Index: 13,02875

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4350 to 1.4390 (20°C, 589 nm)
GC ≥98.0 %
Chloride (Cl) ≤3 ppm
Sulfate (SO4) ≤5 ppm
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR