Diethyl ether 99+% stabilized, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
12399-0010
12399-0025
12399-0050
123990100
123990250
TS12399-0010EA
74
USD
TS12399-0010
TS12399-0025
TS12399-0050
TS123990100
TS123990250
Diethyl ether 99+% stabilized, pure
Diethyl ether
Formula:
(CH₃CH₂)₂O MW: 74.12 g/mol Boiling Pt: 34.6 °C (760 mmHg) Melting Pt: –116 °C Density: 0.714 g/cm³ Flash Pt: –45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I REACH: 01-2119535785-29 Merck Index: 12,03852 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3510 to 1.3540 (20°C, 589 nm) |
Water | ≤0.2 % (Coulometric) |
GC | ≥99.0 % |
Color scale | ≤10 APHA |
Stabilizer | 5 to 10 ppm BHT |
Residue after evaporation | ≤0.002 % |
Peroxides | ≤0.001 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...