Triethylamine 99%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(Diethylamino)ethane, TEA
15791-0010
15791-0025
TS15791-0010EA
154
USD
TS15791-0010
TS15791-0025
TS15791-1000
Triethylamine 99%, pure
Triethylamine
Formula:
(C₂H₅)₃N MW: 101.19 g/mol Boiling Pt: 90 °C (760 mmHg) Melting Pt: –115 °C Density: 0.728 g/cm³ Flash Pt: –11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009051 CAS Number: 121-44-8 EINECS: 204-469-4 UN: 1296 ADR: 3,II Merck Index: 13,09740 |
|
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3990 to 1.4010 (20°C, 589 nm) |
Water | ≤0.2 % (K.F.) |
GC | ≥98.5 % |
Color scale | ≤15 APHA |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...