Iodomethane 99% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl iodide

45901-1000 AC45901-0025 AC45901-5000
TS45901-1000EA 65 USD
TS45901-1000 TS45901-0025 TS45901-5000
Iodomethane 99% stabilised
Iodomethane
Formula: CH₃I
MW: 141.94 g/mol
Boiling Pt: 42.4
Melting Pt: -66
Density: 2.28 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00001073
CAS Number: 74-88-4
UN: 2644
ADR: 6.1 I
Merck Index: 12,06161

Order Now

Specification Test Results

Appearance Clear yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5290 to 1.5310 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Copper


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR