Iodomethane 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
12237-1000
12237-5000
122370025
122370050
TS12237-1000EA
76
USD
TS12237-1000
TS12237-5000
TS122370025
TS122370050
Iodomethane 99% stabilized
Iodomethane
Formula:
CH₃I Boiling Pt: 42.5 °C (760 mmHg) Melting Pt: –66 °C Density: 2.28 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00001073 CAS Number: 74-88-4 EINECS: 200-819-5 UN: 2644 ADR: 6.1,I Merck Index: 12,06161 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5290 to 1.5310 (20°C, 589 nm) |
Water | ≤0.1 % |
GC | ≥98.5 % |
Color scale | ≤150 APHA |
Stabilizer | Copper |
Non-volatile matter | ≤0.01 % |
Heavy metals (as Pb) | ≤0.002 % |
Hydrolyzable substance | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...