Fluoroacetone 97%

Supplier: Thermo Scientific Chemicals
Danger

15178-0050 15178-0010
TS15178-0050EA 260 USD
TS15178-0050 TS15178-0010
Fluoroacetone 97%
Fluoroacetone
Formula: C₃H₅FO
MW: 76.07 g/mol
Boiling Pt: 75 °C (760 mmHg)
Density: 1.054 g/cm³
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000451
CAS Number: 430-51-3
EINECS: 207-064-0
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3660 to 1.3700 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR