Chloroform 99.8% Stabilised analysis_for
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
Stabilization:
Amylen
38377-0025
383770010
383770250
TS38377-0025EA
162
USD
TS38377-0025
TS383770010
TS383770250
Chloroform 99.8% Stabilised analysis_for
Chloroform
Formula:
CHCl₃ MW: 119.38 g/mol Boiling Pt: 61.7 Melting Pt: -63 Density: 1.48 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00000826 CAS Number: 67-66-3 EINECS: 200-663-8 UN: 1888 ADR: 6.1 III REACH: 05-2114614906-45 Merck Index: 13,02160 |
|
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.02 % (K.F.) |
GC | ≥99.8 % |
Color scale | ≤5 APHA |
Stabilizer | Approx. 100 ppm Amylene |
Residue after evaporation | ≤5 ppm |
Acidity/alkalinity | ≤0.0005 meq/g |
Suitability | (for use with Dithizone) Passes test |
Substance darkened by H2SO4 | ≤5 APHA |
Free chlorine | ≤5 ppm |
Calcium (Ca) | ≤0.1 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.02 ppm |
Magnesium (Mg) | ≤0.05 ppm |
Potassium (K) | ≤0.1 ppm |
Sodium (Na) | ≤0.2 ppm |
Zinc (Zn) | ≤0.05 ppm |
Impurity | ≤0.005 % Trichloroethylene |
Total phosphorus (P) | ≤0.1 ppm |
Total silicon (Si) | ≤0.05 ppm |
Total sulfur (S) | ≤0.5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...