Chloroform 99.9% Stabilised, ECD tested Residual, Pest_Residual residue_0.0005
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
Stabilization:
Amylen
32667-0025
326670010
TS32667-0025EA
177
USD
TS32667-0025
TS326670010
Chloroform 99.9% Stabilised, ECD tested Residual, Pest_Residual residue_0.0005
Chloroform
Formula:
CHCl₃ MW: 119.38 g/mol Boiling Pt: 61.7 Melting Pt: -63 Density: 1.48 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00000826 CAS Number: 67-66-3 EINECS: 200-663-8 UN: 1888 ADR: 6.1 III REACH: 05-2114614906-45 Merck Index: 13,02160 |
|
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.01 % (Coulometric) |
GC | ≥99.9 % |
Color scale | ≤10 APHA |
Stabilizer | 0.0035 to 0.005 %w/w Amylene |
GC/ECD (as lindane) | ≤5 ng/l |
Residue after evaporation | ≤0.0005 % |
Acidity (HCl) | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...