Dichloromethane 99.6% stabilised ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methylene chloride

Stabilization: Stabilised with amylene

40692-0040 40692-0250 406920010 406920025 406925000
TS40692-0040EA 206 USD
TS40692-0040 TS40692-0250 TS406920010 TS406920025 TS406925000
Dichloromethane 99.6% stabilised ACS
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8
Melting Pt: -95
Density: 1.325 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.02 % (Coulometric)
GC ≥99.5 %
Color scale ≤10 APHA
Stabilizer 20 to 60 ppm amylene
Residue after evaporation ≤0.002 %
Free halogens passes test
Titrable acid ≤0.0003 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR