Potassium acetate, anhydrous 97%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic acid potassium salt

14960-0025 AC14960-0010 AC14960-5000
TS14960-0025EA 129 USD
TS14960-0025 TS14960-0010 TS14960-5000
Potassium acetate, anhydrous 97%, pure
Potassium acetate
Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 292 °C
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline or granular powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %
Enantiomeric excess ≥98.0 %
Specific optical rotation +15° to +21° (20°C, 589 nm) (c=0.05 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR