Propionic acid 93%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Propanoic acid
14930-0010
14930-0025
14930-0100
TS14930-0010EA
52
USD
TS14930-0010
TS14930-0025
TS14930-0100
Propionic acid 93%, pure
Propionic acid
Formula:
C₃H₆O₂ MW: 74.08 g/mol Boiling Pt: 155 °C (1013 hPa) Melting Pt: –41.5 °C Density: 0.81 g/cm³ Flash Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002756 CAS Number: 79-09-4 EINECS: 209-651-7 UN: 1987 ADR: 8,III REACH: 01-2119486971-24 Merck Index: 13,07917 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4175 to 1.4215 (20°C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤10 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...