Acetic acid 99.8% for biochemistry

Supplier: Thermo Scientific Chemicals
Danger

14893-0010 148930025
TS14893-0010EA 111 USD
TS14893-0010 TS148930025
Acetic acid 99.8% for biochemistry
Acetic acid
Formula: H₃CCOOH
Boiling Pt: 117…118 °C (760 mmHg)
Melting Pt: 16…16.5 °C
Density: 1.048 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
UV at 280 nm A: ≤0.02
Color scale ≤10 APHA
Titration with NaOH ≥99.7 %
Dilution test Passes test
Chloride (Cl) ≤1 ppm
Sulfate (SO4) ≤1 ppm
Cadmium (Cd) ≤0.1 ppm
Iron (Fe) ≤0.2 ppm
Nickel (Ni) ≤0.1 ppm
Heavy metals (as Pb) ≤5 ppm
KMnO4-reducing substance Passes test
K2Cr2O7-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR