Acetic acid 99.7+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
42322-0025
42322-5000
423220100
TS42322-0025EA
177
USD
TS42322-0025
TS42322-5000
TS423220100
Acetic acid 99.7+% ACS
Acetic acid
Formula:
H₃CCOOH Boiling Pt: 118 °C (1013 hPa) Melting Pt: 16…16.5 °C Density: 1.048 g/cm³ Flash Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8,II REACH: 01-2119475328-30 |
|
Specification Test Results
Appearance | Clear liquid |
GC | ≤0.01 % Acetic anhydride ((CH3CO)2O) |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Dilution test | Passes test |
Chloride (Cl) | ≤1 ppm |
Sulfate (SO4) | ≤1 ppm |
Iron (Fe) | ≤0.2 ppm |
Heavy metals (as Pb) | ≤0.5 ppm |
Titrable base | ≤0.0004 meq/g |
KMnO4-reducing substance | Passes test |
K2Cr2O7-reducing substance | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...