Amyl acetate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14918-0010
149180025
149182500
TS14918-0010EA
205
USD
TS14918-0010
TS149180025
TS149182500
Amyl acetate 99%
Amyl acetate
Formula:
C₇H₁₄O₂ Boiling Pt: 149 °C (760 mmHg) Melting Pt: –70.8 °C Density: 0.87 g/cm³ Flash Pt: 24 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009500 CAS Number: 628-63-7 EINECS: 211-047-3 UN: 1104 ADR: 3,III |
|
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4010 to 1.4030 (20°C, 589 nm) |
GC | ≥98.5 % |
TLC | ≥97.5 % |
Specific optical rotation | -10.8° to -11.8° (20°C, 589 nm) (c=2 in DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...