Methyl acetate, extra dry 99+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid methyl ester

37183-1000 37183-0010 371830025
TS37183-1000EA 55 USD
TS37183-1000 TS37183-0010 TS371830025
Methyl acetate, extra dry 99+%, AcroSeal™
Methyl acetate
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 57.4 °C (760 mmHg)
Melting Pt: –98 °C
Density: 0.93 g/cm³
Flash Pt: –10 °C
Storage Temperature: Ambient
MDL Number: MFCD00008711
CAS Number: 79-20-9
EINECS: 201-185-2
UN: 1231
ADR: 3,II
Merck Index: 13,06038

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR