Methyl acetate 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid methyl ester

18138-0010 181380025 181380100
TS18138-0010EA 90 USD
TS18138-0010 TS181380025 TS181380100
Methyl acetate 99%, Extra Pure
Methyl acetate
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 56.9 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.93 g/cm³
Flash Pt: –10 °C
Storage Temperature: Ambient
MDL Number: MFCD00008711
CAS Number: 79-20-9
EINECS: 201-185-2
UN: 1231
ADR: 3,II
Merck Index: 13,06038

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3600 to 1.3620 (20°C, 589 nm)
Water ≤0.05 % (K.F.)
GC ≥98.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Methanol ≤0.7 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR