Ethyl acetate, extra dry 99.9%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid ethyl ester

32690-0010 32690-1000 326900025
TS32690-0010EA 90 USD
TS32690-0010 TS32690-1000 TS326900025
Ethyl acetate, extra dry 99.9%, AcroSeal™
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: -83
Density: 0.902 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.85 %
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR