Isopropyl acetate 99+%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
18080-0025
180800010
180800250
TS18080-0025EA
228
USD
TS18080-0025
TS180800010
TS180800250
Isopropyl acetate 99+%, pure
Isopropyl acetate
Formula:
C₅H₁₀O₂ Boiling Pt: 88.8 °C (760 mmHg) Melting Pt: –73 °C Density: 0.872 g/cm³ Flash Pt: 4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008877 CAS Number: 108-21-4 EINECS: 203-561-1 UN: 1220 ADR: 3,II Merck Index: 13,05225 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...