2-Hexanone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl butyl ketone, Butyl methyl ketone
14688-5000
14688-0025
14688-1000
TS14688-5000EA
347
USD
TS14688-5000
TS14688-0025
TS14688-1000
2-Hexanone 98%
2-Hexanone
Formula:
C₆H₁₂O Boiling Pt: 127 °C (760 mmHg) Melting Pt: –57 °C Density: 0.81 g/cm³ Flash Pt: 23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009482 CAS Number: 591-78-6 EINECS: 209-731-1 UN: 1224 ADR: 3,III Merck Index: 13,06060 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3995 to 1.4015 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...