Acetonitrile 99.8% for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

26827-0010 26827-0040 268270025
TS26827-0010EA 215 USD
TS26827-0010 TS26827-0040 TS268270025
Acetonitrile 99.8% for HPLC
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.781 g/cm³
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 %
GC ≥99.8 %
UV 1 cm cell vs HPLC-grade water
Color scale ≤10 APHA
Residue after evaporation ≤0.0002 %
Acidity ≤0.0008 meq/g
Filtered to 0.2 micrometer Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR