Acetaldehyde 99.5%, AcroSeal™, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethanal

42717-1000 42717-8000
TS42717-1000EA 71 USD
TS42717-1000 TS42717-8000
Acetaldehyde 99.5%, AcroSeal™, Extra Pure
Acetaldehyde
Formula: CH₃CHO
MW: 44.05 g/mol
Boiling Pt: 21 °C (760 mmHg)
Melting Pt: –123 °C
Density: 0.785 g/cm³
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006991
CAS Number: 75-07-0
UN: 1089
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de03379d171eda3c733a039cd16293e8ee4a02e2&vid=573943501bd6753f30bb12e41521b99361988998&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.02 %
GC ≥99.4 %
Stabilizer Approx. 100 ppm Amylene
Residue after evaporation ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR