Acetaldehyde 99.5%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethanal
14951-0010
14951-0100
14951-2500
14951-0025
TS14951-0010EA
250
USD
TS14951-0010
TS14951-0100
TS14951-2500
TS14951-0025
Acetaldehyde 99.5%, Extra Pure
Acetaldehyde
Formula:
CH₃CHO MW: 44.05 g/mol Boiling Pt: 21 °C (760 mmHg) Melting Pt: –123 °C Density: 0.785 g/cm³ Flash Pt: –27 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006991 CAS Number: 75-07-0 EINECS: 200-836-8 UN: 1089 ADR: 3,I Merck Index: 12,00037 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3310 to 1.3330 (20°C, 589 nm) |
Water | ≤0.02 % |
GC | ≥99.4 % |
Chlorine (Cl) | ≤0.003 % (total) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...