1-Bromobutanone 90% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-2-butanone

22245-0250 22245-0010
TS22245-0250EA 764 USD
TS22245-0250 TS22245-0010
1-Bromobutanone 90% stabilized
1-Bromobutanone
Formula: BrH₂CC(O)CH₂CH₃
MW: 151 g/mol
Boiling Pt: 105 °C (150 mmHg)
Density: 1.479 g/cm³
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000207
CAS Number: 816-40-0
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4650 to 1.4690 (20°C, 589 nm)
GC ≥88 %
Stabilizer Approx. 0.5 % CaCO3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR