Lanthanum(III) nitrate hexahydrate 99.995% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
19328-1000
AC19328-0050
AC19328-5000
TS19328-1000EA
270
USD
TS19328-1000
TS19328-0050
TS19328-5000
Lanthanum(III) nitrate hexahydrate 99.995% (trace metals basis)
Lanthanum(III) nitrate hexahydrate
Formula:
La(NO₃)₃·6H₂O MW: 433.01 g/mol Boiling Pt: 126 Melting Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149751 CAS Number: 10277-43-7 EINECS: 233-238-0 UN: 1477 ADR: 5.1 II Merck Index: 14,05363 |
Specification Test Results
Appearance | White Crystals and/or chunks |
Total trace metal impurities | ≤0.005 % |
Lanthanum (La) | 31.0 to 35.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...