Lanthanum(III) chloride heptahydrate 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

19327-1000 AC19327-5000
TS19327-1000EA 298 USD
TS19327-1000 TS19327-5000
Lanthanum(III) chloride heptahydrate 99.99% (trace metals basis)
Lanthanum(III) chloride heptahydrate
Formula: LaCl₃·7H₂O
MW: 371.37 g/mol
Boiling Pt: 860 °C (1013 hPa)
Melting Pt: 91 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0
EINECS: 233-237-5
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Total conc. of imp.as REO ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR