Tin(IV) chloride pentahydrate 98+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tin tetrachloride pentahydrate, Stannic chloride pentahydrate

22369-5000 AC22369-1000
TS22369-5000EA 186 USD
TS22369-5000 TS22369-1000
Tin(IV) chloride pentahydrate 98+%, extra pure
Tin(IV) chloride pentahydrate
Formula: Cl₄Sn·5H₂O
MW: 350.6 g/mol
Melting Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00149864
CAS Number: 10026-06-9
EINECS: 231-588-9
UN: 2440
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals and/or lumps
Assay ≥98.0 %
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR