Tin(II) chloride dihydrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Stannous chloride dihydrate

42448-0025 42448-5000 AC42448-0050 AC42448-1000
TS42448-0025EA 516 USD
TS42448-0025 TS42448-5000 TS42448-0050 TS42448-1000
Tin(II) chloride dihydrate 98+% ACS
Tin (II) chloride dihydrate
Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 37…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 13,08861

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder, crystals and chunks
Titration Iodimetric 98.0 to 103.0 %
Solubility (in hydrochloric acid) Passes test
Sulfate (SO4) Passes test (limit about 0.003%)
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤30 ppm
Lead (Pb) ≤100 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR