Tin(II) chloride dihydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

19680-5000 AC19680-0025 AC19680-0050
TS19680-5000EA 163 USD
TS19680-5000 TS19680-0025 TS19680-0050
Tin(II) chloride dihydrate 97%
Tin(II) chloride dihydrate
Formula: SnCl₂·2H₂O
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 37…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 13,08861

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals and/or chunks
Titration Iodimetric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR