(±)-2-Butanol 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol

10770-0025 107700010 107700100
TS10770-0025EA 141 USD
TS10770-0025 TS107700010 TS107700100
(±)-2-Butanol 99%, Extra Pure
(±)-2-Butanol
Formula: H₃CCH₂CH(OH)CH₃
MW: 74.12 g/mol
Boiling Pt: 99 °C (760 mmHg)
Melting Pt: –115 °C
Density: 0.81 g/cm³
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00004569
CAS Number: 78-92-2
EINECS: 201-158-5
UN: 1120
ADR: 3,III
Merck Index: 13,01540
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dea3607167631fbfcf7751c20b71f345ad24c19&vid=010e04e8f508fd3d501873d20c2349817834b6e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.3980 (20°C, 589 nm)
Water ≤0.5 %
GC ≥98.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR