Methanol-[D₁] 99.0 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

29913-1000 299130250
TS29913-1000EA 199 USD
TS29913-1000 TS299130250
Methanol-[D₁] 99.0 atom % D for NMR spectroscopy
Methanol-[D₁]
Formula: CH₃DO
Boiling Pt: 64.5 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00044213
CAS Number: 1455-13-6
EINECS: 215-933-0
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.1 % (Coulometric)
D-Enrichment ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR