Methanol-[D₃] 99.5 atom % D for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
30073-0100
300730025
TS30073-0100EA
344
USD
TS30073-0100
TS300730025
Methanol-[D₃] 99.5 atom % D for NMR spectroscopy
Methanol-[D₃]
Formula:
D₃COH Boiling Pt: 65 °C (760 mmHg) Density: 0.867 g/cm³ Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055877 CAS Number: 1849-29-2 EINECS: 217-435-9 UN: 1230 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.1 % (Coulometric) |
D-Enrichment | ≥99.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...