Nitromethane-[D₃] (99% D) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

16637-0250 166370050 166370100
TS16637-0250EA 241 USD
TS16637-0250 TS166370050 TS166370100
Nitromethane-[D₃] (99% D) for NMR spectroscopy
Nitromethane-[D₃]
Formula: D₃CNO₂
Boiling Pt: 100…102 °C (760 mmHg)
Density: 1.18 g/cm³
MDL Number: MFCD00044214
CAS Number: 13031-32-8
EINECS: 235-892-2
UN: 1261
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 % (K.F.)
D-Enrichment ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR