Iodomethane[D₃] (99% D) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
17495-0250
174950050
TS17495-0250EA
204
USD
TS17495-0250
TS174950050
Iodomethane[D₃] (99% D) for NMR spectroscopy
Iodomethane[D₃]
Formula:
D₃CI Boiling Pt: 42 °C (1013 hPa) Density: 2.329 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00001074 CAS Number: 865-50-9 EINECS: 212-744-5 UN: 2644 ADR: 6.1,I |
Specification Test Results
Appearance | Clear colorless Liquid (May darken on storage) |
Water | ≤0.05 % (Coulometric) |
D-Enrichment | ≥99.0 % |
HPLC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...