Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Warning
|
32127-0100
321270050
TS32127-0100EA
221
USD
TS32127-0100
TS321270050
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Dichloromethane-[D₂]
Formula:
CD₂Cl₂ Boiling Pt: 40 °C (1013 hPa) Melting Pt: –97 °C Density: 1.36 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000882 CAS Number: 1665-00-5 EINECS: 216-776-0 UN: 1593 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.02 % (Coulometric) |
D-Enrichment | ≥99.75 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...