Dichloromethane-[D₂] 99.5 atom % D for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Warning
|
17611-0250
17611-0100
176110050
176110500
TS17611-0250EA
590
USD
TS17611-0250
TS17611-0100
TS176110050
TS176110500
Dichloromethane-[D₂] 99.5 atom % D for NMR spectroscopy
Dichloromethane-[D₂]
Formula:
CD₂Cl₂ Boiling Pt: 40 °C (1013 hPa) Melting Pt: –97 °C Density: 1.36 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000882 CAS Number: 1665-00-5 EINECS: 216-776-0 UN: 1593 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.04 % (Coulometric) |
D-Enrichment | ≥99.4 % |
Identification C | Passes test |
Identification D | Passes test |
Alkalinity | Passes test |
Titration | ≥85 % |
NaCl and Na2SO4 | ≤8 % |
Non-esterified alcohols | ≤4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...