1,1,2,2-Tetrachloroethane-[D₂] (99.5% D) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,1,2,2-Tetrachloroethane-[²H₂]

32131-0250 32131-0050
TS32131-0250EA 229 USD
TS32131-0250 TS32131-0050
1,1,2,2-Tetrachloroethane-[D₂] (99.5% D) for NMR spectroscopy
1,1,2,2-Tetrachloroethane-[D₂]
Formula: Cl₂CDCDCl₂
MW: 169.83 g/mol
Boiling Pt: 146 °C (760 mmHg)
Density: 1.61 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00037672
CAS Number: 33685-54-0
EINECS: 251-634-1
UN: 1702
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 %
D-Enrichment ≥99.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR