Copper 99.9% (trace metals basis), shot 1-10 mm

Supplier: Thermo Scientific Chemicals

20769-0025 207691000 207695000
TS20769-0025EA 307 USD
TS20769-0025 TS207691000 TS207695000
Copper 99.9% (trace metals basis), shot 1-10 mm
Copper
Formula: Cu
Boiling Pt: 2595 °C (760 mmHg)
Melting Pt: 1083 °C
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Specification Test Results

Appearance Rust brown Metallic shots
Particle size 1 to 10 mm
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR