Tellurium 99.8% (trace metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Warning

31599-5000 315990250 315991000
TS31599-5000EA 515 USD
TS31599-5000 TS315990250 TS315991000
Tellurium 99.8% (trace metals basis), powder -200 mesh
Tellurium
Formula: Te
MW: 127.6 g/mol
Boiling Pt: 990 °C (760 mmHg)
Melting Pt: 450 °C
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
Merck Index: 12,09273

Order Now

Specification Test Results

Appearance Gray to black Powder
Total trace metal impurities ≤0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR