Lithium, dry 99+%, granular

Supplier: Thermo Scientific Chemicals
Danger

21144-0250 211442500
TS21144-0250EA 76 USD
TS21144-0250 TS211442500
Lithium, dry 99+%, granular
Lithium
Formula: Li
MW: 6.94 g/mol
Boiling Pt: 1317 °C (1013 hPa)
Melting Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00134051
CAS Number: 7439-93-2
EINECS: 231-102-5
UN: 1415
ADR: 4.3 I
Merck Index: 13,05542

Order Now

Specification Test Results

Appearance Silver gray Granulate
Assay ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR