Aluminium oxide 99.99% (trace metals basis), extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

AC21225-0250 AC21225-1000
TS21225-0250EA 101 USD
TS21225-0250 TS21225-1000
Aluminium oxide 99.99% (trace metals basis), extra pure
Aluminium oxide
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2977 °C (760 mmHg)
Melting Pt: 2030 °C
Density: 3.97 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now

Specification Test Results

Appearance White Powder
Total trace metal impurities ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR