Tetramethylammonium acetate hydrate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetramethylammonium acetate xhydrate
AC42049-0050
AC42049-0250
TS42049-0050EA
48
USD
TS42049-0050
TS42049-0250
Tetramethylammonium acetate hydrate 95%
Tetramethylammonium acetate hydrate
Formula:
C₆H₁₅NO₂·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD03066126 CAS Number: 1266615-86-4 EINECS: 234-179-3 |
Specification Test Results
Appearance | White Crystalline solid |
Infrared spectrum | Conforms |
Water | ≤14 % (K.F.) |
Titration with HClO4 | ≥94 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...