Tetraethylammonium chloride hydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrylammonium chloride hydrate

AC15090-0250 AC15090-1000 AC15090-5000
TS15090-0250EA 53 USD
TS15090-0250 TS15090-1000 TS15090-5000
Tetraethylammonium chloride hydrate 99%
Tetraethylammonium chloride hydrate
Formula: C₈H₂₀ClN·xH₂O
MW: 165.71 g/mol (anhy)
MDL Number: MFCD00149992
CAS Number: 56-34-8
EINECS: 200-267-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 % (on dry substance)
Loss on drying ≤10 %
Impurity ≤0.5 % Triethylamine hydrochloride

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR