Sodium metatungstate in water stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium polytungstate, hexa-Sodium tungstate (W12)

AC30410-0500 AC30410-2500
TS30410-0500EA 354 USD
TS30410-0500 TS30410-2500
Sodium metatungstate in water stabilized
Sodium metatungstate
Formula: H₂Na₆O₄₀W₁₂
MW: 2986.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150663
CAS Number: 12141-67-2

Order Now

Specification Test Results

Appearance Clear yellow to light green Liquid
Assay ≥67 % (WO3)
Stabilizer Typically 0.02 % Na2Cr2O7 salt

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR