Ammonium iron(III) oxalate trihydrate 98%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC33510-0010
AC33510-0051
AC33510-0500
TS33510-0010EA
60
USD
TS33510-0010
TS33510-0051
TS33510-0500
Ammonium iron(III) oxalate trihydrate 98%, pure
Ammonium iron(III) oxalate trihydrate
Formula:
(NH₄)₃Fe(C₂O₄)₃·3H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150137 CAS Number: 13268-42-3 UN: 2923 ADR: 8,III |
Specification Test Results
Appearance | Green Crystalline powder |
Infrared spectrum | Conforms |
Titration with KMnO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...