Magnesium carbonate heavy heavy, extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesium carbonate hydroxide, Magnesium carbonate basic, heavy

AC41340-0025 AC41340-0100 AC41340-5000
TS41340-0025EA 210 USD
TS41340-0025 TS41340-0100 TS41340-5000
Magnesium carbonate heavy heavy, extra pure
Magnesium carbonate heavy
Formula: CH₂Mg₂O₅
MW: 142.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011107
CAS Number: 39409-82-0
EINECS: 208-915-9

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric 40 to 45 % (MgO)
Loss on calcination 55 % to 60 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR