(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (R)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole

AC36978-0050 AC36978-0250
TS36978-0050EA 89 USD
TS36978-0050 TS36978-0250
(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
(R)-(+)-2-Methyl-CBS-oxazaborolidine
Formula: C₁₈H₂₀BNO
MW: 277.17 g/mol
Boiling Pt: 111 °C (760 mmHg)
Density: 0.95 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00078440
CAS Number: 112022-83-0
UN: 1294
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to amber Solution
Strength 0.90 to 1.10 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR