Bis(benzonitrile)palladium(II) dichloride 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bis(benzonitrile)palladium chloride, Dichlorobis(benzonitrile)palladium(II), Bis(benzonitrile)palladium(II) Dichloride
AC20790-0010
AC20790-0100
TS20790-0010EA
90
USD
TS20790-0010
TS20790-0100
TS20790-2500
Bis(benzonitrile)palladium(II) dichloride 97+%
Bis(benzonitrile)dichloropalladium(II)
Formula:
C₁₄H₁₀Cl₂N₂Pd Melting Pt: 120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013123 CAS Number: 14220-64-5 EINECS: 238-085-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Yellow to orange to brown Powder |
Infrared spectrum | Conforms |
Elemental Analysis | ≥97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...